Bis(cyclopentadienyl)dimethylhafnium ≥97%

Supplier: STREM CHEMICALS INC MS

72-0700-2G 72-0700-500MG
100203-262EA 660.98 USD
100203-262 100203-260
Bis(cyclopentadienyl)dimethylhafnium ≥97%
Bis(cyclopentadienyl)dimethylhafnium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₂H₁₆Hf
Boiling Pt: 90 °C (0.1 mmHg) (subl.)
MDL Number: MFCD02093015
CAS Number: 37260-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR