Ytterbium(III) trifluoromethanesulphonate hydrate

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Trifluoromethanesulphonic acid ytterbium salt hydrate, Ytterbium(III) trifluoromethanesulphonate hydrate, Ytterbium(III) triflate hydrate, Ytterbium triflate hydrate

70-6000-5G 70-6000-1G 70-6000-25G
100207-732EA 83.51 USD
100207-732 100207-728 100207-730
Ytterbium(III) trifluoromethanesulphonate hydrate
Ytterbium(III) trifluoromethanesulphonate hydrate

Metal Triflate

Formula: C₃F₉O₉S₃Yb·xH₂O
MW: 620.25 g/mol (anhy)
MDL Number: MFCD03703499
CAS Number: 252976-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR