Ytterbium(III) hexafluoroacetylacetonate dihydrate ≥99.9% (Yb basis)

Supplier: STREM CHEMICALS INC MS
Warning

70-2500-5G 70-2500-1G
100204-640EA 714.77 USD
100204-640 100198-236
Ytterbium(III) hexafluoroacetylacetonate dihydrate ≥99.9% (Yb basis)
Ytterbium(III) hexafluoroacetylacetonate dihydrate

Metal Beta-diketonates, Volatile Precursors for CVD

MDL Number: MFCD00167115
CAS Number: 81849-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR