Ytterbium(III) 2,4-pentanedionate hydrate ≥99.9% (Yb basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ytterbium(III) acetylacetonate hydrate

70-2202-25G 70-2202-5G
100207-426EA 568.98 USD
100207-426 100199-432
Ytterbium(III) 2,4-pentanedionate hydrate ≥99.9% (Yb basis)
Ytterbium(III) 2,4-pentanedionate hydrate
Formula: C₁₅H₂₁O₆Yb·xH₂O
MW: 470.37 g/mol (anhy)
MDL Number: MFCD00013508
CAS Number: 14284-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR