Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ytterbium(III) ≥99% (99.9%-Yb)

Supplier: STREM CHEMICALS INC MS

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III), Yb(TMHD)3

70-0100-1G 70-0100-5G
100210-040EA 58.03 USD
100210-040 100196-328
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ytterbium(III) ≥99% (99.9%-Yb)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ytterbium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇O₆Yb
MW: 722.842 g/mol
Boiling Pt: 300 °C (dec.)
Melting Pt: 167…169 °C
MDL Number: MFCD00000034
CAS Number: 15492-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR