Tris(cyclopentadienyl)ytterbium(III) ≥99.9% (Yb basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tris(cyclopentadienyl)ytterbium

70-0075-5G 70-0075-1G
100203-804EA 625.6 USD
100203-804 100199-430
Tris(cyclopentadienyl)ytterbium(III) ≥99.9% (Yb basis)
Tris(cyclopentadienyl)ytterbium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₅H₁₅Yb
MW: 368.32 g/mol
Boiling Pt: 150 °C (subl. in vacuum)
Melting Pt: 273 °C
MDL Number: MFCD00064762
CAS Number: 1295-20-1
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR