Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')thulium(III) ≥98% (99.9%-Tm)

Supplier: STREM CHEMICALS INC MS

Synonyms: Tm(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)thulium(III)

69-7000-1G 69-7000-5G
100211-154EA 89.17 USD
100211-154 100198-870
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')thulium(III) ≥98% (99.9%-Tm)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')thulium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₃₃H₅₇O₆Tm
MW: 718.736 g/mol
Boiling Pt: 280 °C (dec.)
Melting Pt: 169…172 °C
MDL Number: MFCD00015712
CAS Number: 15631-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR