Tris(cyclopentadienyl)thulium ≥99.9% (Tm basis)

Supplier: STREM CHEMICALS INC MS
Warning

69-6000-5G 69-6000-1G
100196-882EA 839.32 USD
100196-882 100200-292
Tris(cyclopentadienyl)thulium ≥99.9% (Tm basis)
Tris(cyclopentadienyl)thulium

Metallocenes, Derivatives & Cp Precursors, Volatile Precursors for CVD

MDL Number: MFCD01631285
CAS Number: 1272-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR