Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')erbium(III) ≥99% (99.9%-Er)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)erbium(III), Er(TMHD)3
68-8750-1G
68-8750-25G
68-8750-5G
100205-184EA
66.52
USD
100205-184
100205-186
100205-188
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')erbium(III) ≥99% (99.9%-Er)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')erbium(III)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formula:
C₃₃H₅₇ErO₆ Boiling Pt: 345 °C/ 160 °C (0.1 mmHg) (dec./ subl.) Melting Pt: 179…180 °C |
MDL Number:
MFCD00015705 CAS Number: 35733-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...