Tris(methylcyclopentadienyl)erbium(III) ≥99.9% (Er basis)

Supplier: STREM CHEMICALS INC MS
Warning

68-8740-1G 68-8740-5G
100207-724EA 175.51 USD
100207-724 100207-726
Tris(methylcyclopentadienyl)erbium(III) ≥99.9% (Er basis)
Tris(methylcyclopentadienyl)erbium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

MDL Number: MFCD03427338
CAS Number: 39470-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR