Tris(cyclopentadienyl)erbium(III) ≥99.9%

Supplier: STREM CHEMICALS INC MS
Danger

68-8000-1G 68-8000-5G
100208-044EA 144.37 USD
100208-044 100202-680
Tris(cyclopentadienyl)erbium(III) ≥99.9%
Tris(cyclopentadienyl)erbium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₅H₁₅Er
MW: 362.539 g/mol
Boiling Pt: 200 °C (0,01 mmHg) (subl.)
Melting Pt: 285 °C
MDL Number: MFCD00145475
CAS Number: 39330-74-0
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR