Tris(isopropylcyclopentadienyl)erbium(III) ≥99.9%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tris(i-propylcyclopentadienyl)erbium(III)

68-7200-5G 68-7200-1G
100200-290EA 672.31 USD
100200-290 100211-152
Tris(isopropylcyclopentadienyl)erbium(III) ≥99.9%
Tris(isopropylcyclopentadienyl)erbium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₂₄H₃₃Er
MW: 488.778 g/mol
Boiling Pt: 222 °C (10 mmHg) (subl.)
Melting Pt: 222 °C (subl.)
MDL Number: MFCD00145481
CAS Number: 130521-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR