Tris(n-butylcyclopentadienyl)erbium(III) ≥99.9%

Supplier: STREM CHEMICALS INC MS

68-7000-1G 68-7000-5G
100211-148EA 208.06 USD
100211-148 100211-150
Tris(n-butylcyclopentadienyl)erbium(III) ≥99.9%
Tris(n-butylcyclopentadienyl)erbium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Boiling Pt: 240 °C (0.1 mmHg)
Density: 1.309
MDL Number: MFCD00145486
CAS Number: 153608-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR