Erbium(III) hexafluoroacetylacetonate hydrate ≥99.9%

Supplier: STREM CHEMICALS INC MS
Warning

68-6900-5G 68-6900-1G
100203-254EA 962.46 USD
100203-254 100203-252
Erbium(III) hexafluoroacetylacetonate hydrate ≥99.9%
Erbium(III) hexafluoroacetylacetonate hydrate

Metal Beta-diketonates, Volatile Precursors for CVD

MDL Number: MFCD03427337
CAS Number: 18923-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR