Erbium(III) trifluoromethanesulfonate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
68-6000-5G
68-6000-25G
101442-368EA
100.49
USD
101442-368
101442-366
Erbium(III) trifluoromethanesulfonate ≥98%
Erbium(III) trifluoromethanesulphonate
Metal Triflate
Formula:
C₃ErF₉O₉S₃ |
MDL Number:
MFCD00192126 CAS Number: 139177-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...