Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')dysprosium(III) ≥98% (99.9%-Dy)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Dy(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)dysprosium(III)
66-8500-1G
66-8500-5G
100197-570EA
66.52
USD
100197-570
100197-572
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')dysprosium(III) ≥98% (99.9%-Dy)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')dysprosium(III)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formula:
C₃₃H₅₇DyO₆ MW: 712.302 g/mol Boiling Pt: 265 °C (dec.) Melting Pt: 182…185 °C |
MDL Number:
MFCD00000017 CAS Number: 15522-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...