Tris(isopropylcyclopentadienyl)dysprosium(III) ≥99.9% (Dy basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
66-3000-5G
66-3000-1G
101442-486EA
774.22
USD
101442-486
101442-484
Tris(isopropylcyclopentadienyl)dysprosium(III) ≥99.9% (Dy basis)
Tris(isopropylcyclopentadienyl)dysprosium(III)
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD
Formula:
C₂₄H₃₃Dy |
MDL Number:
MFCD10566478 CAS Number: 952518-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...