Dysprosium(III) 2,4-pentanedionate hydrate ≥99.9% (dye basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dysprosium (III) acetylacetonate hydrate

66-2002-5G 66-2002-25G 66-2002-100G
100196-326EA 110.4 USD
100196-326 100210-036 100198-868
Dysprosium(III) 2,4-pentanedionate hydrate ≥99.9% (dye basis)
Dysprosium (III) acetylacetonate hydrate
Formula: C₁₅H₂₁DyO₆· xH₂O
MW: 459.83 g/mol (anhy)
MDL Number: MFCD00013489
CAS Number: 14637-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR