Tris(isopropylcyclopentadienyl)terbium(III) ≥99.9% (Tb basis)

Supplier: STREM CHEMICALS INC MS
Warning

65-7000-5G 65-7000-1G
100203-250EA 1006.34 USD
100203-250 100198-028
Tris(isopropylcyclopentadienyl)terbium(III) ≥99.9% (Tb basis)
Tris(isopropylcyclopentadienyl)terbium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₂₄H₃₃Tb
MDL Number: MFCD00798537
CAS Number: 312696-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR