Tris(tetramethylcyclopentadienyl)gadolinium(III) ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

64-4500-1G 64-4500-5G
100207-424EA 452.92 USD
100207-424 100208-042
Tris(tetramethylcyclopentadienyl)gadolinium(III) ≥98%
Tris(tetramethylcyclopentadienyl)gadolinium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Precursors for CVD

MDL Number: MFCD01862470
CAS Number: 308847-85-0
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR