Tris(cyclopentadienyl)gadolinium(III) ≥99.9% (Gd basis)

Supplier: STREM CHEMICALS INC MS
Danger

64-4000-5G 64-4000-1G
100205-974EA 751.57 USD
100205-974 100201-720
Tris(cyclopentadienyl)gadolinium(III) ≥99.9% (Gd basis)
Tris(cyclopentadienyl)gadolinium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₅H₁₅Gd
MW: 352.535 g/mol
MDL Number: MFCD00145476
CAS Number: 1272-21-5
UN: 1325
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR