Gadolinium(III) isopropoxide ≥99.9% (Gd basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Gadolinium(III) i-propoxide

64-3700-1G 64-3700-5G
100201-012EA 158.52 USD
100201-012 100201-014
Gadolinium(III) isopropoxide ≥99.9% (Gd basis)
Gadolinium(III) isopropoxide

Alkoxide

Formula: C₉H₂₁GdO₃
MW: 334.514 g/mol
MDL Number: MFCD00144381
CAS Number: 14532-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR