Gadolinium(III) hydroxide hydrate ≥99.9% (Gd basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
64-3600-10G
64-3600-50G
100199-426EA
158.52
USD
100199-426
100207-422
Gadolinium(III) hydroxide hydrate ≥99.9% (Gd basis)
Gadolinium(III) hydroxide hydrate
Formula:
Gd(OH)₃·xH₂O MW: 208.27 g/mol (anhy) |
MDL Number:
MFCD00049930 CAS Number: 100634-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...