Gadolinium(III) 2-ethylhexanoate ∼25% in toluene

Supplier: STREM CHEMICALS INC MS
Danger

64-3500-50G 64-3500-10G
100201-718EA 396.31 USD
100201-718 100201-716
Gadolinium(III) 2-ethylhexanoate ∼25% in toluene
Gadolinium(III) 2-ethylhexanoate
MDL Number: MFCD00269820
CAS Number: 19189-19-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR