Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')samarium(III) ≥99.9% (Sm basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Sm(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)samarium(III)
62-4000-5G
62-4000-1G
100199-254EA
328.37
USD
100199-254
100200-486
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')samarium(III) ≥99.9% (Sm basis)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')samarium(III)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formula:
C₃₃H₅₇O₆Sm Boiling Pt: 250 °C/ 183 °C (1.3 mmHg) (dec./ subl.) Melting Pt: 191…193 °C |
MDL Number:
MFCD00015711 CAS Number: 15492-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...