Tris(cyclopentadienyl)samarium(III) ≥99.9% (Sm basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tris(cyclopentadienyl)samarium

62-3500-5G 62-3500-1G
100206-124EA 788.37 USD
100206-124 100198-234
Tris(cyclopentadienyl)samarium(III) ≥99.9% (Sm basis)
Tris(cyclopentadienyl)samarium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₅H₁₅Sm
MW: 345.64 g/mol
Melting Pt: 356 °C
MDL Number: MFCD00013755
CAS Number: 1298-55-1
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR