Samarium(III) trifluoromethanesulphonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Samarium(III) trifluoromethanesulphonate

62-3300-25G 62-3300-5G
101442-464EA 358.09 USD
101442-464 101442-466
Samarium(III) trifluoromethanesulphonate ≥98%
Samarium(III) trifluoromethanesulphonate

Metal Triflate

Formula: C₃F₉O₉S₃Sm
MW: 597.57 g/mol
MDL Number: MFCD00192431
CAS Number: 52093-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR