Samarium diiodide 0.1M in THF

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Samarium (II) iodide

62-3100-250ML 62-3100-50ML
100196-322EA 257.6 USD
100196-322 100196-324
Samarium diiodide 0.1M in THF
Samarium diiodide
Formula: SmI₂
MW: 404.17 g/mol
Density: 0.922 g/mL
MDL Number: MFCD00058873
CAS Number: 32248-43-4
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR