Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')neodymium(III) ≥99% (99.9%-Nd)

Supplier: STREM CHEMICALS INC MS

Synonyms: Nd(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)neodymium(III)

60-8750-5G 60-8750-1G
100197-162EA 329.78 USD
100197-162 100207-716
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')neodymium(III) ≥99% (99.9%-Nd)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')neodymium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇NdO₆
MW: 694.044 g/mol
Boiling Pt: 270 °C/ 150 °C (0.1 mmHg)
Melting Pt: 209…212 °C (dec./ subl.)
MDL Number: MFCD00015708
CAS Number: 15492-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR