Tris(tetramethylcyclopentadienyl)neodymium(III) ≥99.9% (Nd basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tris(tetramethylcyclopentadienyl)neodymium

60-7500-5G 60-7500-1G
100208-040EA 996.43 USD
100208-040 100207-420
Tris(tetramethylcyclopentadienyl)neodymium(III) ≥99.9% (Nd basis)
Tris(tetramethylcyclopentadienyl)neodymium(III)

Metallocenes, Derivatives & Cp Precursors

Formula: C₂₇H₃₉Nd
MW: 507.841 g/mol
MDL Number: MFCD01862468
CAS Number: 164528-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR