Tris(isopropylcyclopentadienyl)neodymium(III) ≥99.9% (Nd basis)

Supplier: STREM CHEMICALS INC MS
Warning

60-6000-5G 60-6000-1G
100206-782EA 890.28 USD
100206-782 100210-822
Tris(isopropylcyclopentadienyl)neodymium(III) ≥99.9% (Nd basis)
Tris(isopropylcyclopentadienyl)neodymium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₂₄H₃₃Nd
MDL Number: MFCD00145483
CAS Number: 69021-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR