Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')neodymium(III) ≥99% (99.9%-Nd)

Supplier: STREM CHEMICALS INC MS

Synonyms: Nd(FOD)3, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)neodymium(III)

60-5500-1G 60-5500-5G
100200-710EA 121.72 USD
100200-710 100208-458
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')neodymium(III) ≥99% (99.9%-Nd)
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')neodymium(III)
Formula: C₃₀H₃₀F₂₁NdO₆
MW: 1029.764 g/mol
MDL Number: MFCD00798419
CAS Number: 17978-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR