Neodymium(III) trifluoromethanesulphonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Neodymium triflate, Neodymium(III) trifluoromethanesulphonate

60-5200-25G 60-5200-5G
101442-414EA 324.12 USD
101442-414 101442-416
Neodymium(III) trifluoromethanesulphonate ≥98%
Neodymium(III) trifluoromethanesulphonate

Metal Triflate

Formula: C₃F₉NdO₉S₃
MW: 591.45
MDL Number: MFCD00192343
CAS Number: 34622-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR