Tris(cyclopentadienyl)neodymium(III) ≥99% (99.9%-Nd)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
60-5000-5G
60-5000-1G
60-5000-25G
100199-252EA
338.28
USD
100199-252
100201-714
100205-972
Tris(cyclopentadienyl)neodymium(III) ≥99% (99.9%-Nd)
Tris(cyclopentadienyl)neodymium(III)
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD
Formula:
C₁₅H₁₅Nd Boiling Pt: 220 °C (0.01 mmHg) (subl.) |
MDL Number:
MFCD00145477 CAS Number: 1273-98-9 UN: 1325 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...