Tris(isopropylcyclopentadienyl)praseodymium(III) ≥99.9% (Pr basis)

Supplier: STREM CHEMICALS INC MS
Warning

59-8000-1G 59-8000-5G
100211-146EA 182.58 USD
100211-146 100200-288
Tris(isopropylcyclopentadienyl)praseodymium(III) ≥99.9% (Pr basis)
Tris(isopropylcyclopentadienyl)praseodymium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₂₄H₃₃Pr
MDL Number: MFCD00145484
CAS Number: 69021-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR