Praseodymium(III) isopropoxide ≥99.9% (Pr basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Praseodymium(III) i-propoxide

59-5000-5G 59-5000-1G
100206-778EA 433.11 USD
100206-778 100206-776
Praseodymium(III) isopropoxide ≥99.9% (Pr basis)
Praseodymium (III) isopropoxide

Alkoxide

Formula: C₉H₂₁O₃Pr
MW: 318.169 g/mol
Boiling Pt: 175 °C (0.04 mmHg) (subl.)
MDL Number: MFCD00145157
CAS Number: 19236-14-7
UN: 3178
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR