Praseodymium(III) acetylacetonate hydrate ≥99.9% (Pr basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Praseodymium(III) acetylacetonate hydrate, Praseodymium 2,4-Pentanedionate, Praseodymium acetylacetonate hydrate
59-2002-5G
59-2002-100G
59-2002-25G
100196-318EA
66.52
USD
100196-318
100203-244
100198-230
Praseodymium(III) acetylacetonate hydrate ≥99.9% (Pr basis)
Praseodymium(III) acetylacetonate hydrate
Formula:
C₁₅H₂₆O₇Pr MW: 459.274 g/mol |
MDL Number:
MFCD08272322 CAS Number: 14553-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...