Tris(tetramethylcyclopentadienyl)cerium(III) ≥99.9% (Ce basis)

Supplier: STREM CHEMICALS INC MS
Warning

58-9000-1G 58-9000-5G
100209-372EA 210.89 USD
100209-372 100209-374
Tris(tetramethylcyclopentadienyl)cerium(III) ≥99.9% (Ce basis)
Tris(tetramethylcyclopentadienyl)cerium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₂₇H₃₉Ce
MDL Number: MFCD01863662
CAS Number: 251984-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR