Tris(i-propylcyclopentadienyl)cerium(III) ≥99.9% (Ce basis)

Supplier: STREM CHEMICALS INC MS
Warning

58-8000-5G 58-8000-1G
100202-678EA 1139.38 USD
100202-678 100205-844
Tris(i-propylcyclopentadienyl)cerium(III) ≥99.9% (Ce basis)
Tris(i-propylcyclopentadienyl)cerium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

MDL Number: MFCD00269815
CAS Number: 122528-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR