Tris(cyclopentadienyl)cerium(III) ≥99.9% (Ce basis)

Supplier: STREM CHEMICALS INC MS

58-7500-5G 58-7500-1G
100207-264EA 404.8 USD
100207-264 100203-242
Tris(cyclopentadienyl)cerium(III) ≥99.9% (Ce basis)
Tris(cyclopentadienyl)cerium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Boiling Pt: 230 °C (0.01 mmHg) (subl.)
Melting Pt: 452 °C (dec.)
MDL Number: MFCD00145474
CAS Number: 1298-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR