Cerium(III) stearate, Technical Grade

Supplier: STREM CHEMICALS INC MS

58-1000-2500G 58-1000-500G
100203-240EA 450.09 USD
100203-240 100205-180
Cerium(III) stearate, Technical Grade
Cerium(III) stearate
Melting Pt: 120…124 °C
MDL Number: MFCD00070456
CAS Number: 10119-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR