Tris(isopropylcyclopentadienyl)lanthanum(III) ≥99.9% (La basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Tris(i-propylcyclopentadienyl)lanthanum
57-4000-5G
57-4000-1G
100203-238EA
666.65
USD
100203-238
100203-236
Tris(isopropylcyclopentadienyl)lanthanum(III) ≥99.9% (La basis)
Tris(isopropylcyclopentadienyl)lanthanum(III)
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD
Formula:
C₂₄H₃₃La MW: 460.43 g/mol Boiling Pt: 180…195 °C (0.02 mmHg) |
MDL Number:
MFCD00145482 CAS Number: 68959-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...