Tris(cyclopentadienyl)lanthanum(III) ≥99.9% (La basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Tris(cyclopentadienyl)lanthanum

57-3000-1G 57-3000-5G
100196-314EA 128.8 USD
100196-314 100208-676
Tris(cyclopentadienyl)lanthanum(III) ≥99.9% (La basis)
Tris(cyclopentadienyl)lanthanum(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₅H₁₅La
MW: 334.19 g/mol
Melting Pt: 295 °C (dec.)
MDL Number: MFCD00045467
CAS Number: 1272-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR