Lanthanum(III) trifluoromethanesulfonate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Lanthanum(III) trifluoromethanesulphonate, Lanthanum trifluoromethanesulphonate, Lanthanum triflate

57-2800-2G 57-2800-10G
100198-862EA 41.05 USD
100198-862 100196-312
Lanthanum(III) trifluoromethanesulfonate ≥97%
Lanthanum(III) trifluoromethanesulphonate

Metal Triflate

Formula: C₃F₉LaO₉S₃
MW: 586.12 g/mol
MDL Number: MFCD00077560
CAS Number: 52093-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR