Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lanthanum(III) ≥99% (99.9%-La)
Supplier: STREM CHEMICALS INC MS
Synonyms:
La(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III)
57-1000-5G
57-1000-25G
57-1000-1G
100205-968EA
290.15
USD
100205-968
100205-966
100207-714
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lanthanum(III) ≥99% (99.9%-La)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lanthanum(III)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formula:
C₃₃H₅₇LaO₆ MW: 688.71 g/mol Boiling Pt: 370 °C/ 210 °C (0.2 mmHg) (dec./ subl.) Melting Pt: 227…231 °C |
MDL Number:
MFCD00010466 CAS Number: 14319-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...