Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium triglyme adduct ≥99.99% (Ba basis) (Sr-0.5%), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

56-8610-5G 56-8610-1G
100197-026EA 1759.32 USD
100197-026 100210-820
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium triglyme adduct ≥99.99% (Ba basis) (Sr-0.5%), PURATREM
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium triglyme adduct

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Melting Pt: 88 °C
MDL Number: MFCD01073800
CAS Number: 149160-45-2
UN: 1564
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR