Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium tetraglyme adduct ≥99.99% (Ba basis) (Sr-0.5%), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
56-8600-5G
56-8600-1G
100195-920EA
1520.12
USD
100195-920
100195-918
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium tetraglyme adduct ≥99.99% (Ba basis) (Sr-0.5%), PURATREM
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium tetraglyme adduct
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Melting Pt:
103 °C |
MDL Number:
MFCD01073799 CAS Number: 136629-60-2 UN: 1564 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...