Barium trifluoromethanesulfonate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Barium Bis(trifluoromethanesulphonate), Barium triflate, Barium(II) trifluoromethanesulphonate, Trifluoromethanesulphonic acid Barium(II) salt, Ba(OTf)2, Barium(II) triflate

56-8100-25G 56-8100-5G
100197-564EA 278.83 USD
100197-564 100207-712
Barium trifluoromethanesulfonate ≥99%
Barium trifluoromethanesulphonate

Metal Triflate

Formula: C₂BaF₆O₆S₂
MW: 435.45 g/mol
Melting Pt: Dec.
MDL Number: MFCD00143644
CAS Number: 2794-60-7
UN: 1564
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR