Barium isopropoxide, dried powder

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Barium i-propoxide

56-8000-25G 56-8000-5G
100208-938EA 706.28 USD
100208-938 100208-940
Barium isopropoxide, dried powder
Barium isopropoxide

Alkoxide

Formula: C₆H₁₄BaO₂
MW: 255.501 g/mol
MDL Number: MFCD00050486
CAS Number: 24363-37-9
UN: 3205
ADR: 4.2,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR