Barium neodecanoate (25 - 29% Ba), Superconductor Grade

Supplier: STREM CHEMICALS INC MS
Warning

56-7000-10G 56-7000-50G
100210-014EA 120.31 USD
100210-014 100210-016
Barium neodecanoate (25 - 29% Ba), Superconductor Grade
Barium neodecanoate
MDL Number: MFCD00070454
CAS Number: 55172-98-0
UN: 1564
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR